ContactUsLinkComponent
Attest™ Rapid Readout Biological Indicators, 3M™
Attest™ Rapid Readout Biological Indicators, 3M™
Catalog # CA34010-532
Supplier:  Avantor
Attest™ Rapid Readout Biological Indicators, 3M™
Catalog # CA34010-532
Supplier:  Avantor
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Rapid Readout Biological Indicators (Brown cap) for 250°F (121°C) Gravity and 270°F (132°C) Vacuum Assisted Steam Sterilizers, Three-hour Readout
  • Cat. no.:
    CA34010-532
  • Supplier No.:
    1292
Specifications
About this item
Attest™ Rapid readout self-contained biological indicator is a dual readout system specifically designed for rapid and reliable monitoring of steam sterilization process.
  • Color-coded for quick reference and easy use
  • Reduce possibility of contamination and minimizes false positives
  • Faster result time, making it possible to quarantine every load and eliminate recalls
These indicators detect the presence of Geobacillus stearothermophilus by detecting the activity of alpha‑glucosidase, an enzyme present within the organism. The presence of the enzyme is detected by reading fluorescence produced by the enzymatic breakdown of a non‑fluorescent substrate. This creates a fluorescence change, which is detected by the auto‑reader. A fluorescence change indicates a steam sterilization process failure.