ContactUsLinkComponent
Aura™ 9211+ N95 Particulate Respirators, 3M™
Aura™ 9211+ N95 Particulate Respirators, 3M™
Catalog # CA10031-466
Supplier:  3M
undefined
Aura™ 9211+ N95 Particulate Respirators, 3M™
Catalog # CA10031-466
Supplier:  3M
Supplier Number:  7100110432
Specifications
  • Color:
    White
  • Description:
    Flat Fold Particulate Respirator with CoolFlow™ Valve
  • Cat. no.:
    CA10031-466
  • Supplier No.:
    9211+
Specifications
About this item
This three-panel disposable flat fold respirator features 3M™ Cool Flow™ Valve to help provide comfortable, reliable worker respiratory protection.
  • Lightweight, 3-panel, flat fold, disposable
  • Protection against non-oil based particles
Features a braided headbands provide comfort and help minimize pulling of hair
These are used in Grinding, Sanding, Sweeping, Bagging and other dusty or arid operations. Also in other areas such as Textile Operations, Dry Chemical Handling, Surface Preparation, Food Preparation and Handling, Woodworking, Grain Handling and Milling, Processing of Minerals, and certain other substances.
Caution: These respirators help reduce exposures to certain airborne particulates. Before use, the wearer must read and understand the User Instructions provided as a part of the product packaging. A written respiratory protection program must be implemented meeting all the requirements of OSHA 1910.134 including training, fit testing and medical evaluation.