Orders LinkContactUsLinkComponent
Biox Cryogenic Storage Boxes
Biox Cryogenic Storage Boxes
Supplier:   TWD Scientific
Biox Cryogenic Storage Boxes
Supplier:  TWD Scientific
Certificates
Biox Cryogenic Storage Boxes
Biox Cryogenic Storage Boxes
2 Options Available Below
Product Details & Documents
Cryogenic storage boxes are manufactured from polycarbonate for liquid nitrogen and deep freezer use.
  • Clear removable lid
  • Numbered grip
  • Drain holes
  • 1 and 2" options available
Always in a 10×10 configuration. Openings in the bottom make them scan ready when used with 2D barcoded vials.
Ordering information: Storage boxes are packaged 12 per case.
Product Family Options
Product Information
  • No. of Holes
    Dimensions
    Material
    Availability
    Price
  • 100
    133L×133W×53H mm
    Polycarbonate
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $419.40
    $419.40
    Case of 12
       $419.40
    Supplier Number: BX98-0213
  • 100
    133L×133W×36H mm
    Polycarbonate
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $409.86
    $409.86
    Case of 12
       $409.86
    Supplier Number: BX98-0113