If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Primary antibody reactivity:Human
- Sensitivity:97 pg/ml (range 0.1 - 3.2 ng/ml)
- Cat. no.:89141-790
- Supplier No.:960-076
- No. of Tests:96 wells
Specifications
About this item
Most comprehensive portfolio of heat shock protein (chaperone) products commercially available.
For the quantitative determination of human HSP27 in cell lysates, plasma, and serum.
The ImmunoSet® HSP27 high sensitivity (human), EIA development set contains sufficient reagents for the development of 5 x 96 well EIA plates which provides analysis of ~200 samples in duplicate. Save time, money, and sample by developing your own EIA kits. Simply coat your plates and go! This kit provides superior accuracy with fully quantitative results compared to Western blot analysis and has low reactivity with related molecules.