PGE2 High Sensitivity ELISA Kit, Enzo Life Sciences
89141-766
:
- Assay Type:Competitive
- Target protein:PGE2
- Sensitivity:8.26 pg/mL (range 7.8 - 1,000 pg/mL)
- Cat. no.:89141-766
- Supplier No.:931-001
- No. of Tests:96 wells
Ultrasensitive PGE2 ELISA kit for inflammation and eicosanoid research.
For the quantitative determination of PGE2 in culture supernatants, serum, saliva, urine, and whole blood from any species.
The PGE2 high sensitivity EIA kit is a colorimetric competitive enzyme immunoassay kit with results overnight + 1 hour.
: Contains GxM IgG Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-762 (1x96 wells).