Order Entry
Canada
ContactUsLinkComponent
ImmunoSet® αB-Crystallin ELISA development set, Enzo Life Sciences
ImmunoSet® αB-Crystallin ELISA development set, Enzo Life Sciences
Catalog # 89141-786
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
ImmunoSet® αB-Crystallin ELISA development set, Enzo Life Sciences
Catalog # 89141-786
Supplier:  Enzo Life Sciences
Supplier Number:  ADI-960-074
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Assay duration:
    Multiple steps
  • Format:
    Uncoated
  • Primary antibody reactivity:
    Human
    Bovine
    Rat
    Mouse
  • Target protein:
    αB-Crystallin
  • Description:
    ImmunoSet® αB-Crystallin ELISA development set
  • Size:
    5 × 96 wells
  • Detection method:
    Colorimetric 450 nm
  • Sensitivity:
    0.59 ng/ml (range 1.25 - 40 ng/ml)
  • Cat. no.:
    89141-786
  • Supplier No.:
    960-074
  • No. of Tests:
    96 wells

Specifications

About this item

For the quantitative determination of αB-Crystallin in cell lysates of human, mouse, rat, and cow origin.

The ImmunoSet® αB-Crystallin EIA development set contains sufficient reagents for the development of 5 x 96 well EIA plates which provides analysis of ~200 samples in duplicate. Save time, money, and sample by developing your own EIA kits. Simply coat your plates and go! This kit provides superior accuracy with fully quantitative results compared to Western blot analysis and has low reactivity with related molecules.

Ordering information: Contains Capture antibody, Standard, Detection antibody, SA-HRP.