ContactUsLinkComponent
PGE2 FPIA Kit, Enzo Life Sciences
PGE2 FPIA Kit, Enzo Life Sciences
  89141-756
 :  
undefined
PGE2 FPIA Kit, Enzo Life Sciences
  89141-756
 :  ADI-920-001
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
  • Sensitivity:
    684 pg/mL (range 1,562 - 100,000 pg/mL)
  • Cat. no.:
    89141-756
  • Supplier No.:
    920-001
  • No. of Tests:
    100
For the quantitative determination of PGE2 in culture supernatants from any species.
The PGE2 FPIA kit is a homogeneous mix-and-read fluorescence polarization enzyme immunoassay kit with. Results are obtained within just 30 minutes and the generated signal is stable for up to 20 hours. The PGE2 FPIA kit is ideal for high throughput screening (HTS) applications. A convenient homogeneous mix-and-read assay that is amendable to miniaturization and automation, this kit has a robust Z factor >0.8.
 : Contains Conjugate concentrate, Antibody, Assay buffer concentrate, Standard.