If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Sensitivity:(Nitrite) 0.222 µmole/L; (Nitrate) 0.625 µmole/L (range (Nitrite) 3.125 - 200 µmole/L; (Nitrate) 3.125 - 100 µmole/L)
- Cat. no.:89141-752
- Supplier No.:917-010
- No. of Tests:96 wells
For the quantitative determination of nitrite and nitrate levels in culture supernatants, plasma, serum, saliva, and urine from any species.
The Nitric Oxide (NO2-/NO3-), detection kit is a complete kit for the quantitative determination of nitrate and nitrite concentrations. Absorbance is read at 540-570 nm. Only 0.050 mL diluted sample needed per well saves precious sample.