Leptin (Mouse), ELISA Kit, Enzo Life Sciences
Catalog # 89141-076
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Primary antibody reactivity:Mouse
- Target protein:Leptin
- Sensitivity:25.4pg/mL (range 50 - 3,200pg/mL)
- Cat. no.:89141-076
- Supplier No.:900-019A
- No. of Tests:96 wells
Specifications
About this item
For the quantitative determination of mouse leptin in culture supernatants, plasma, and serum
The Leptin (mouse), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours. Absorbance is read at 450nm. The broad dynamic range is suitable for many normal and pathological samples with sensitive quantification of < 26pg/ml mouse leptin.
Cited sample type includes tissue.
Ordering information: Contains Microtiter plate, Standard, Antibody, Conjugate, Assay buffer 28, TMB Substrate, Stop solution 2, Wash buffer concentrate.