PGE2 ELISA Kit, Enzo Life Sciences
Catalog # 89141-052
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Assay Type:Competitive
- Conjugate ELISA:Alkaline Phosphatase (AP)
- Format:Pre-coated
- Primary antibody reactivity:Species independent
- Target protein:PGE2
- Description:PGE2 ELISA kit
- Detection method:Colorimetric detection 405 nm
- Time to Results:<3 hours
- Sensitivity:13.4 pg/ml (range 39.1 - 2,500 pg/ml)
- Cat. no.:89141-052
- Supplier No.:900-001
- No. of Tests:96 wells
Specifications
About this item
Highly sensitive PGE2 ELISA kit for inflammation and eicosanoid research.
For the quantitative determination of PGE2 in culture supernatants, serum, saliva, urine, and whole blood from any species. Cited sample types include cell lysate, cerebral spinal fluid, dialysate, gingival crevicular fluid, peritoneal exudate fluid and peritoneal exudate cell supernatant, plasma, and tissue.
The PGE2 EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 2. Commercially available since 1992, this kit is widely cited in peer-reviewed publications. The non-radioactive ready-to-use liquid color-coded reagents reduce errors.
Ordering information: Contains GxM IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-268 (5x96 wells).
Caution: Store all components at +4 °C, except standard and conjugate at –20 °C.