Anti-Streptavidin Rabbit Polyclonal Antibody (Biotin)
Catalog # CA200-406-095S
Supplier: Rockland Immunochemical
Specifications
- Antibody type:Tag
- Antigen name:Streptavidin
- Antigen symbol:Streptavidin
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Size:25 µL
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin. Immunoglobulin and Protease free.
- Concentration:1.0mg/ml by UV absorbance at 280 nm
- Tested applications:Fluoresence ELISA
- Cat. no.:CA200-406-095S
- Supplier No.:200-406-095S
Specifications
About this item
Biotin Conjugated Anti-Streptavidin Antibody has been assayed by ELISA for the detection of streptavidin in a standard ELISA using Peroxidase as a reporter.
Recommended Dilutions: ELISA: 1:2000-1:20000;
Type: Tag
Antigen: Streptavidin
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: