Specifications
- Cat. no.:89156-274
- Supplier No.:2814-0500
Specifications
About this item
The Opioid and Sigma Ligands Plate contains 74 ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs.
The library is ideal for screening or identifying recombinant orphan G protein coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The Opioid and Sigma Ligands plate is 1 plate of the larger Neurotransmitter Library which contains over 650 ligands.