Order Entry
Canada
ContactUsLinkComponent
Chemiluminescent Labeling Kit, Enzo Life Sciences
Chemiluminescent Labeling Kit, Enzo Life Sciences
Catalog # 89141-662
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Chemiluminescent Labeling Kit, Enzo Life Sciences
Catalog # 89141-662
Supplier:  Enzo Life Sciences
Supplier Number:  ADI-907-001
CAS Number:  

Specifications

  • Host:
  • Description:
    Chemiluminescent Labeling Kit
  • Environmentally Preferable:
  • Cat. no.:
    89141-662
  • No. of Reactions:
    5
  • Supplier No.:
    907-001

Specifications

About this item

A complete kit for the labeling of antibodies, proteins, peptides, and nucleic acids with Acridinium NHS Ester.

Chemiluminescent acridinium esters are commonly used to label antibodies, proteins, nucleic acids, and peptides containing primary amine groups. These chemiluminescent molecules are frequently for use as detection reagents in immunoassays as a safe alternative to radioactive tracers. The Chemiluminescent labeling kit provides the necessary components for quick, efficient, and easy acridinium labeling.

Ordering information: Contains Acridinium NHS Ester, Dry DMF, Bicarbonate labeling buffer, 10% Lysine, Gel filtration column, Column buffer concentrate, Set of Trigger solutions.