Order Entry
Canada
Orders LinkContactUsLinkComponent
Acridinium Ester, (Trigger Solution), Enzo Life Sciences

Acridinium Ester, (Trigger Solution), Enzo Life Sciences

250 mL
Catalog # 89141-658
Supplier:   Enzo Life Sciences
Supplier #:  ADI-906-001
Acridinium Ester, (Trigger Solution), Enzo Life Sciences
Catalog # 89141-658
Supplier:  Enzo Life Sciences
CAS Number:  
Acridinium Ester, (Trigger Solution), Enzo Life Sciences

250 mL

CAS Number:  
Price
$306.52
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Species Independent

Liquid. Solution 1 contains hydrogen peroxide in nitric acid. Solution 2 contains cetyl trimethylammonium chloride in sodium hydroxide.

Specifications

Specifications

  • Size:
    250 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.