PGF2α ELISA Kit, Enzo Life Sciences
Catalog # 89141-316
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Assay Type:Competitive
- Target protein:PGF2α
- Sensitivity:6.71 pg/mL (range 3.05 - 50,000 pg/mL)
- Cat. no.:89141-316
- Supplier No.:901-069
- No. of Tests:96 wells
Specifications
About this item
For the quantitative determination of PGF2α in culture supernatants, plasma, serum, saliva, and urine from any species
The PGF2α EIA kit is a colorimetric competitive enzyme immunoassay kit with results in < 3 hours. Absorbance is read at 405 nm. Broad dynamic range is suitable for a wide variety of sample types. Ready-to-use liquid color-coded reagents are safe, non-radioactive, and reduce laboratory errors.
Cited sample types include cell lysate and whole blood.
Ordering information: Contains DxS Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-120 (1x96 wells).