ContactUsLinkComponent
Shipper, Multisample EHS Ambient, Therapak®
  10029-400
undefined
Shipper, Multisample EHS Ambient, Therapak®
  10029-400
 :  56875
  • Description:
    Large transport box, multi-sample shipper
  • Cat. no.:
    10029-400
  • Supplier No.:
    56875
Large capacity ambient shipping system accommodates multiple specimen transport bags for consolidation at the collection site and shipment to testing laboratory
System includes a large transport box (ID: 11 x 10.5 x 4"; OD: 11.25 x 10.75 x 4.25") with easy lock flaps on both ends of the box, a 12 x 18" biohazard transport bag, 12 x 12" absorbent sheet. Holds up to ten 6 x 9" specimen transport bags.