Order Entry
Canada
ContactUsLinkComponent
PGE2 ELISA Kit, Enzo Life Sciences
PGE2 ELISA Kit, Enzo Life Sciences
  89141-268
 :  
undefined
PGE2 ELISA Kit, Enzo Life Sciences
  89141-268
 :  ADI-901-001
 :  

 

  • Assay Type:
    Competitive
  • Conjugate ELISA:
    Alkaline Phosphatase (AP)
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Species independent
  • Target protein:
    PGE2
  • Description:
    PGE2 ELISA kit
  • Detection method:
    Colorimetric detection 405 nm
  • Time to Results:
    <3 hrs
  • Sensitivity:
    13.4 pg/ml (range 39.1 - 2,500 pg/ml)
  • Cat. no.:
    89141-268
  • Supplier No.:
    901-001
  • No. of Tests:
    96 wells

 

 

Highly sensitive PGE2 ELISA kit for inflammation and eicosanoid research.

For the quantitative determination of PGE2 in culture supernatants, serum, saliva, urine, and whole blood from any species. Cited sample types include cell lysate, cerebral spinal fluid, dialysate, gingival crevicular fluid, peritoneal exudate fluid and peritoneal exudate cell supernatant, plasma, and tissue.

The PGE2 EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 2. Commercially available since 1992, this kit is widely cited in peer-reviewed publications. The non-radioactive ready-to-use liquid color-coded reagents reduce errors.

 : Contains GxM Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-052 (1x96 wells).
 : Store all components at +4 °C, except standard and conjugate at –20 °C.