p62 ELISA Kit, Enzo Life Sciences
Catalog # 89409-178
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Sensitivity:100pg/mL (range 625 - 40,000pg/mL)
- Cat. no.:89409-178
- Supplier No.:9002120001
- No. of Tests:96 wells
Specifications
About this item
Highly validated, quantitative p62 ELISA kit available for autophagy research.
For the quantitative determination of human, rat and mouse p62 in cell lysate samples. Cited sample type includes in PBMC lysates.
The p62 ELISA kit is a colorimetric, immunometric immunoassay kit with results in 3 hours.
Ordering information: Contains Microtiter Plate, Antibody, Conjugate, Assay Buffer 13, Wash Buffer Concentrate, Standard, TMB Substrate, Stop Solution 2, RIPA Cell Lysis Buffer 2.