Order Entry
Canada
ContactUsLinkComponent
Residual Hardness Test, MQuant®, MilliporeSigma
Residual Hardness Test, MQuant®, MilliporeSigma
Catalog # CA1.11142.0001
Supplier:  MilliporeSigma
CAS Number:  
Residual Hardness Test, MQuant®, MilliporeSigma
Catalog # CA1.11142.0001
Supplier:  MilliporeSigma
Supplier Number:  1.11142.0001
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Chemical method:
    Mixed indicator
  • Method:
    Colorimetric with color card 0.04 - 0.08 - 0.15 °d 0.05 - 0.1 - 0.19°e 0.7 - 1.4 - 2.7 mg/l CaCO₃
  • Storage temperature:
    Store at +15°C to +25°C.
  • Cat. no.:
    CA1.11142.0001
  • Range:
    0.04 - 0.08 - 0.15 °d 0.05 - 0.1 - 0.19°e 0.7 - 1.4 - 2.7 mg/L CaCO₃
  • Supplier No.:
    1.11142.0001
  • No. of Tests:
    400

Specifications

About this item

The residual hardness is measured semiquantitatively by visual comparison of the colour of the measurement solution with the colour fields of the colour card.