Order Entry
Canada
ContactUsLinkComponent
Phosphate Test, Spectroquant®, MilliporeSigma
Phosphate Test, Spectroquant®, MilliporeSigma
Catalog # CA1.14842.0001
Supplier:  MilliporeSigma
CAS Number:  
Phosphate Test, Spectroquant®, MilliporeSigma
Catalog # CA1.14842.0001
Supplier:  MilliporeSigma
Supplier Number:  1.14842.0001
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Chemical method:
    Vanadate-molybdate
  • Description:
    Phosphate Test
  • Method:
    Colorimetric, VM, with color card and comparator block with short test tubes 1.0 - 2.0 - 3.5 - 6.0 - 10 - 20 - 40 mg/l PO₄-P 3.1 - 6.1 - 11 - 18 - 31 - 61 - 123 mg/l PO₄³⁻
  • Storage temperature:
    Store at +15°C to +25°C.
  • Cat. no.:
    CA1.14842.0001
  • Range:
    1.0 - 2.0 - 3.5 - 6.0 - 10 - 20 - 40 mg/L PO₄-P 3.1 - 6.1 - 11 - 18 - 31 - 61 - 123 mg/L PO₄³⁻
  • Supplier No.:
    1.14842.0001
  • No. of Tests:
    400
  • For Unit:
    Prove, Nova 60, Move

Specifications

About this item

All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments

  • Highly stable, ready to use reagent mixtures in a user friendly design
  • Free selection of appropriate cell format according to the measuring range
  • Long shelf life up to 3 years - all at room temperature
  • Automatic selection of the correct method with help of AutoSelector barcode identification

Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.

*7 This method is officially recognised by the USEPA as an alternative method for the investigation of the following:
*7-1 wastewater
*7-2 drinking water
*7-3 respectively drinking water and waste water

Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).