Fluid Thioglycollate Medium in ReadyRack™, Disposable Tube Rack, Hardy Diagnostics
Catalog # CA10436-340
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Specifications
- Description:Fluid Thioglycollate Medium in ReadyRack™, 16 places
- Cat. no.:CA10436-340
- Supplier No.:K212BX
Specifications
About this item
Fluid Thioglycollate Medium in ReadyRack™ is recommended for the cultivation of aerobic, microaerophilic, and anaerobic microorganisms in normally sterile materials
- 80 Tube Ready Rack
- Holes in the bottom of the rack allow for sterilant to drain
- Convenient hand-holds
- No cardboard for low particle counts
The ReadyRack is a plastic rack designed for cleanroom use and can be rinsed with alcohol. It is a convenient way to bring media into a controlled area while avoiding outside contamination.
Simply remove ReadyRack from box outside of cleanroom. Take into cleanroom and remove plastic bag. Spray down entire ReadyRack with isopropyl alcohol, and allow to dry.
Ordering information: Fluid Thioglycollate Medium in ReadyRack, USP, contains 16 x 125 mm tubes, 10 mL fill. This product must be ordered by the package of 80.