Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Chemical method:Diphenylcarbazide
- Description:Chromate Test
- Method:Colorimetric with color-disk comparator 0.10 - 0.20 - 0.35 - 0.6 - 1.0 - 1.8 - 3.0 - 6.0 - 10 mg/l Cr 0.22 - 0.45 - 0.8 - 1.3 - 2.2 - 4.0 - 6.7 - 13 - 22 mg/l CrO₄²⁻
- Storage temperature:Store at +15°C to +25°C.
- Cat. no.:CA1.14756.0001
- Range:0.10 - 0.20 - 0.35 - 0.6 - 1.0 - 1.8 - 3.0 - 6.0 - 10 mg/L Cr 0.22 - 0.45 - 0.8 - 1.3 - 2.2 - 4.0 - 6.7 - 13 - 22 mg/L CrO₄²⁻
- Supplier No.:1.14756.0001
- No. of Tests:300
Specifications
About this item
The test measures chromium(VI) present in the sample as chromate or dichromate ions in wastewater and percolating water, groundwater and surface water, industrial water and drinking water.
When the reagents of theMColortestTM test are used up, the kit can be replenished by ordering the appropriate refill pack. Refill packs are available for a range of MColortestTM test kits, making it a particularly economical system.