About this item
Greiner Bio-One offers a complete line of low volume tubes with draw volumes of 1 to 2.5 mL to facilitate reduction of phlebotomy volumes, improve overall patient care, and minimize the amount of discarded specimen waste.
- Minimizes discarded specimen waste
- Low level of volume required for samples
- Color-coded safety caps
All low volume tubes are standard 13x75mm plastic tubes with color-coded safety caps. Tubes offer functional compatability with instrumentation.