Nalgene® Syringe Filters, Nylon, 25mm, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
These 25 mm Ø Nylon syringe filters are ideal for filtering samples and mobile phases used in GC and HPLC.
- Sterile filters are certified sterile, non-cytotoxic and non-pyrogenic
- Accommodate sample volumes from 10 to 100 ml
Nylon membrane is naturally hydrophilic, has low extractables and some protein binding. The nylon membrane and polypropylene housing are chemically-resistant to a wide range of solvents. Pink housing ring.
They are also useful for filtering alcohols, cryoprotectants such as DMSO, aqueous/biological solutions and weak solvents.
Packaging: Sterile filters are individually blister-packed. Non sterile filters are bulk packed in zip closure bags.
Customers Who Bought This Also Bought