Evidence Vial Containers with Chain of Custody Closures, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Capitol evidence containers with chain of custody closures feature a one-way, irreversible chain lock that secures the container once it is closed.
- 100% FDA approved food grade polypropylene
- Sterile, airtight and leak-resistant construction with permanent hinged lid
- Shoulderless, linerless containers
- To open after use, simply cut the chain lock with scissors
- Ideal for transport and storage of sensitive materials and samples
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.