KIMTECH PURE® A5 Cleanroom Integrated Hood and Mask, Kimberly-Clark
Catalog #: 10160-684
Supplier: Kimberly-Clark
Waste Reducing
Specifications
- Clothing Size:One size
- Garment application:Cleanroom
- Color:White
- Environmentally Preferable:Waste Reducing
- Cat. no.:10160-684
- Packaging:75/Case
- Supplier No.:49828
Specifications
About this item
In cleanroom environments, people are the number one source of contamination, and the garments they wear can make a big difference
- Breathable SMS fabric
- Clean processed
- Bound, tightly sewn seams offer added protection and strengthen barriers
- Stretch fit elastic
- Combines two donning steps into one
- Pull down ties in back to assist in the gowning process
- Bulk packaged for convenience and waste reduction
- Recyclable through the RightCycle™ by Kimberly-Clark Professional®
The KIMTECH PURE® A5 Cleanroom Integrated Hood and Mask is designed to protect cleanroom processes by providing comfortable and sustainable protection.
Customers Who Bought This Also Bought