Mycoplasma Agar with Cefoperazone, Hardy Diagnostics
Catalog # CA10753-454
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Specifications
- Pack type:Plate
- Description:Mycoplasma Agar with 18 mL Cefoperazone
- Size:15 x 100 mm
- Cat. no.:CA10753-454
- Supplier No.:G212
Specifications
About this item
Mycoplasma agar with cefoperazone is used for the detection of Mycoplasma spp
associated with bovine mastitis.