Orders LinkContactUsLinkComponent
Technical Buffer Solutions, Sachets
Technical Buffer Solutions, Sachets
Technical Buffer Solutions, Sachets
Supplier:  Mettler Toledo
Certificates
Technical Buffer Solutions, Sachets
Technical Buffer Solutions, Sachets
5 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
NIST-traceable buffer for pH electrode calibration and verification.
  • NIST-traceable
  • Ensure freshness
  • Improve data
Also available in multiple packaging options.
Mettler Toledo buffer sachets ensure a fresh portion of buffer is used for each calibration, improving your pH data.
Product Family Options
Product Information
  • pH accuracy
    Pack type
    Size
    pH
    Availability
    Price
  • ±0.02
    Sachet, red
    30×20 ml
    4.01
    Log in for availability
    Log in for availability
    Pack of 30
    Pack of 30 - $169.00
    $169.00
    Pack of 30
       $169.00
    Supplier Number: 51302069
  • Sachet
    30×20 ml
    2.00
    Log in for availability
    Log in for availability
    Log in for availability
    Each(20ml)
    Each(20ml) - $192.96
    $192.96
    New Customer Promotional Price
    Log in for availability
    Pack of 30 (20ml)
    Pack of 30 (20ml) - $183.22
    $183.22
    New Customer Promotional Price
    Each(20ml)
       $192.96
    Pack of 30 (20ml)
       $183.22
    Supplier Number: 30111134
  • ±0.02
    Sachet, green
    30×20 ml
    7.00
    Log in for availability
    Log in for availability
    Pack of 30
    Pack of 30 - $169.00
    $169.00
    Pack of 30
       $169.00
    Supplier Number: 51302047
  • ±0.02
    Sachets, blue
    30×20 ml
    10.00
    Log in for availability
    Log in for availability
    Pack of 30
    Pack of 30 - $169.00
    $169.00
    Pack of 30
       $169.00
    Supplier Number: 51302079
  • ±0.02
    Sachet
    3×10×20 ml
    4.01/7.00/10.01
    Log in for availability
    Log in for availability
    Pack of 30
    Pack of 30 - $169.00
    $169.00
    Pack of 30
       $169.00
    Supplier Number: 51302080
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.