About this item
The Multifuge™ X1 Centrifuge series offers industry-leading throughput, the most versatile range of rotors and adapters, and energy-efficient operation. Recommended for use in cell culture, microplate, bioproduction, food safety, blood separation, bodily fluid processing, pharmaceutical, biotechnology, stem cell, microbiology, molecular biology, proteomics,and genomics applications. This centrifuge series offers unparalleled processing performance, maximum versatility, and exceptional energy efficiency. A broad range of rotors and adapters, including lightweight Fiberlite carbon fiber rotors, are available to handle such diverse laboratory applications.
To facilitate simple operation, each unit features an intuitive control panel and digital display
The Auto-Lock™ III Rotor System is a secure locking system that allows easy push-button installation and exchange of rotors. The system saves time in between applications, and offers the user easy access to the chamber, enabling quick cleaning, a healthier working environment, and longer unit life.
The SMARTSpin™ Technology is an advanced rotor management system which maximizes acceleration, braking and residual load imbalance for each rotor and bucket option. By this technique, system optimizes safety and improves separations.
Detailed operation information, including display of set and actual parameters, maximize run performance and temperature accuracy for improved sample protection. The user can easily adjust speed, time, and temperature, even during the run, with a convenient keyboard for direct input. Innovative lid locking technology enables one finger downward motion for simple closing and locking of the centrifuge lid, even on tall benches (with optional automatic lid opening). A double temperature management system displays the air temperature in the chamber as well as the sample temperature, ensuring accurate control of the sample temperature during the run.