GsBP®-1MS Non-Polar GC Columns, GS-Tek
Supplier: General Separation Technologies, Inc.
Certificates
About this item
Typical Application: Amines, volatile, Diesel / Gasoline, Drug Screen, Ethylene Glycol Mixture, Hydrocarbons Sim-Dis, Pyrolysates of Polystyrene, Steroids: Sex Hormones, Sulfur Compounds.
- 100% poly(methylsiloxane) phase
- Equivalent to USP G1, G2, G38
Ordering information: These columns are suitable replacements for HP-1MS, DB-1MS, CP-Sil 5CB MS, VF-1MS, Rtx-1MS, Rxi-1MS.
Specifications
- Catalog No:
- 10499-094
- 10499-096
- 10499-098
- 10499-130
- 10499-086
- 10499-132
- 10499-088
- 10499-134
- 10499-090
- 10499-092
- 10499-326
- 10499-328
- 10499-330
- 10499-320
- 10499-322
- 10499-324
- 10499-100
- 10499-102
- 10499-104
- 10499-118
- 10499-076
- 10499-078
- 10499-108
- 10499-110
- 10499-112
- 10499-114
- 10510-884
- 10510-886
- 10499-122
- 10499-124
- 10499-080
- 10499-126
- 10499-082
- 10499-128
- 10499-084
- 10499-318
- 10499-120
- Description:
- 10499-094
- 10499-096
- 10499-098
- 10499-130
- 10499-086
- 10499-132
- 10499-088
- 10499-134
- 10499-090
- 10499-092
- 10499-326
- 10499-328
- 10499-330
- 10499-320
- 10499-322
- 10499-324
- 10499-100
- 10499-102
- 10499-104
- 10499-118
- 10499-076
- 10499-078
- 10499-108
- 10499-110
- 10499-112
- 10499-114
- 10510-884
- 10510-886
- 10499-122
- 10499-124
- 10499-080
- 10499-126
- 10499-082
- 10499-128
- 10499-084
- 10499-318
- 10499-120
- Temperature limit:
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C