Anti-ATRAID Rabbit Polyclonal Antibody
10155-686
- Antibody type:Primary
- Antigen name:All-trans Retinoic Acid-induced differentiation Factor
- Antigen symbol:ATRAID
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:150 µL
- Antigen synonyms:ATRAID
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Storage temperature:Store at –20 °C
- Immunogen:ATRAID
- Purification:Antigen affinity purification
- Cat. no.:10155-686
- Supplier No.:25548-1-AP
The ATRAID antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human ATRAID. This antibody recognizes human, mouse antigen. The ATRAID antibody has been validated for the following applications: ELISA, WB analysis.
IHC 1 : 50
Type: Primary
Antigen: ATRAID
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse