Anti-SLC25A22 Rabbit Polyclonal Antibody
10155-604
- Antibody type:Primary
- Antigen name:Solute Carrier Family 25 Member A22
- Antigen symbol:SLC25A22
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:150 µL
- Antigen synonyms:GC1|NET44|EIEE3
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Storage temperature:Store at –20 °C
- Immunogen:SLC25A22
- Purification:Antigen affinity purification
- Cat. no.:10155-604
- Supplier No.:25402-1-AP
The SLC25A22 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human SLC25A22. This antibody recognizes human, mouse, rat antigen. The SLC25A22 antibody has been validated for the following applications: ELISA, WB analysis.
LL
Type: Primary
Antigen: SLC25A22
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat