Anti-Drebrin Rabbit Polyclonal Antibody
Catalog # 10155-778
Supplier: Proteintech
Specifications
- Antibody type:Primary
- Antigen symbol:Drebrin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:150 µL
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Storage temperature:Store at –20 °C
- Immunogen:Drebrin
- Purification:Antigen affinity purification
- Cat. no.:10155-778
- Supplier No.:25770-1-AP
Specifications
About this item
The Drebrin antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human Drebrin. This antibody recognizes human, mouse antigen. The Drebrin antibody has been validated for the following applications: ELISA, WB analysis.
WB 1 : 300
Type: Primary
Antigen: Drebrin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse