To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Size:100 mL
- Storage temperature:Store undiluted at 2-30°C
- Tested applications:Flow Cytometry
- Cat. no.:10274-088
- Supplier No.:TNB-4700-L100
Specifications
About this item
Lymphocyte Separation Medium (LSM) is a sterile-filtered iso-osmotic solution designed for the in vitro isolation of lymphocytes from diluted whole blood
The use of LSM is based on the method of isolating lymphocytes using density gradient centrifugation (differential centrifugation), resulting in the formation of several distinct cell layers. Mononuclear cells (lymphocytes and monocytes) and platelets will be contained in the banded plasma-LSM interphase due to their density. Erythrocytes and granulocytes will migrate through the gradient to the bottom of the tube. Lymphocytes can be recovered by aspirating the plasma layer and removing the cells.