Order Entry
Canada
ContactUsLinkComponent
 
Anti-BTN3A1 Rabbit Polyclonal Antibody
  10155-496
undefined
Anti-BTN3A1 Rabbit Polyclonal Antibody
  10155-496
 :  25221-1-AP-150UL

 

  • Antibody type:
    Primary
  • Antigen name:
    Butyrophilin, subfamily 3, member A1
  • Antigen symbol:
    BTN3A1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    150 µL
  • Antigen synonyms:
    BTF5|BT3.1|BTN3.1|CD277
  • Storage buffer:
    PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
  • Storage temperature:
    Store at –20 °C
  • Immunogen:
    BTN3A1
  • Purification:
    Antigen affinity purification
  • Cat. no.:
    10155-496
  • Supplier No.:
    25221-1-AP

 

 

The BTN3A1 Antibody from Proteintech is a rabbit polyclonal Antibody to a fusion protein of human BTN3A1. This Antibody recognizes human Antigen. The BTN3A1 Antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 700 ; IP 1 : 500

Type: Primary
Antigen: BTN3A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human