Anti-TREX1 Rabbit Polyclonal Antibody
Catalogue # 10155-336
Fournisseur: Proteintech
Spécifications
- Antibody type:Primary
- Antigen name:Three Prime Repair Exonuclease 1
- Antigen symbol:TREX1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:150 µL
- Antigen synonyms:TREX1|HERNS|CRV|Three prime repair exonuclease 1|DRN3|DNase III|AGS1
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Storage temperature:Store at –20 °C
- Immunogen:TREX1
- Purification:Antigen affinity purification
- Cat. no.:10155-336
- Supplier No.:24876-1-AP
Spécifications
À propos de cet article
The TREX1 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human TREX1. This antibody recognizes human antigen. The TREX1 antibody has been validated for the following applications: ELISA, WB analysis.
WB 1 : 1000
Type: Primary
Antigen: TREX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human