Anti-PIK3C2B Rabbit Polyclonal Antibody
Catalog # 10155-312
Supplier: Proteintech
Specifications
- Antibody type:Primary
- Antigen name:Phosphatidylinositol-4-Phosphate 3-Kinase, Catalytic Subunit Type 2 Beta
- Antigen symbol:PIK3C2B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:150 µL
- Antigen synonyms:C2-PI3K
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Storage temperature:Store at –20 °C
- Immunogen:PIK3C2B
- Purification:Antigen affinity purification
- Cat. no.:10155-312
- Supplier No.:24788-1-AP
Specifications
About this item
The PIK3C2B antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human PIK3C2B. This antibody recognizes human antigen. The PIK3C2B antibody has been validated for the following applications: ELISA, WB, IP analysis.
WB 1 : 700 ; IHC 1 : 50
Type: Primary
Antigen: PIK3C2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human