Order Entry
Canada
ContactUsLinkComponent
 
Molecular sieve A4 (0.4 nm, 4 Å), beads ~ 2 mm, with moisture indicator, Supelco®
Catalog # CA1.05739.1000
Supplier:  MilliporeSigma
CAS Number:  
Molecular sieve A4 (0.4 nm, 4 Å), beads ~ 2 mm, with moisture indicator, Supelco®
Catalog # CA1.05739.1000
Supplier:  MilliporeSigma
Supplier Number:  1.05739.1000
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 kg
  • Cat. no.:
    CA1.05739.1000
  • Supplier No.:
    1.05739.1000

Specifications

About this item

 :
CAS Number:70955-01-0
 :1318-02-1
MDL Number:MFCD03457537