Order Entry
Canada
ContactUsLinkComponent
Anti-CREST Mouse Monoclonal Antibody [clone: 3D5D10]
Anti-CREST Mouse Monoclonal Antibody [clone: 3D5D10]
Catalog # 10155-898
Supplier:  Proteintech
undefined
Anti-CREST Mouse Monoclonal Antibody [clone: 3D5D10]
Catalog # 10155-898
Supplier:  Proteintech
Supplier Number:  60314-1-IG-150UL

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    CREST
  • Clonality:
    Monoclonal
  • Clone:
    3D5D10
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    150 µL
  • Storage buffer:
    PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
  • Storage temperature:
    Store at –20 °C
  • Immunogen:
    CREST
  • Purification:
    Protein G purification
  • Cat. no.:
    10155-898
  • Supplier No.:
    60314-1-IG

Specifications

About this item

The CREST antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human CREST. This antibody recognizes human antigen. The CREST antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 2000

Type: Primary
Antigen: CREST
Clonality: Monoclonal
Clone: 3D5D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: