Order Entry
Canada
ContactUsLinkComponent
Anti-F9 Rabbit Polyclonal Antibody
Anti-F9 Rabbit Polyclonal Antibody
Catalog # 10154-956
Supplier:  Proteintech
undefined
Anti-F9 Rabbit Polyclonal Antibody
Catalog # 10154-956
Supplier:  Proteintech
Supplier Number:  21481-1-AP-150UL

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Coagulation Factor 9
  • Antigen symbol:
    F9
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    150 µL
  • Antigen synonyms:
    P19|THPH8|Christmas factor|FIX|F9 p22|HEMB|Coagulation Factor IX|PTC
  • Storage buffer:
    PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
  • Storage temperature:
    Store at –20 °C
  • Immunogen:
    F9
  • Purification:
    Antigen affinity purification
  • Cat. no.:
    10154-956
  • Supplier No.:
    21481-1-AP

Specifications

About this item

The F9 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human F9. This antibody recognizes human antigen. The F9 antibody has been validated for the following applications: ELISA, WB analysis.

IHC 1 : 50

Type: Primary
Antigen: F9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human