Order Entry
ContactUsLinkComponent
Anti-ARPP-21 Rabbit Polyclonal Antibody
Anti-ARPP-21 Rabbit Polyclonal Antibody
Catalog # 10155-988
Supplier:  Proteintech
undefined
Anti-ARPP-21 Rabbit Polyclonal Antibody
Catalog # 10155-988
Supplier:  Proteintech
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    cAMP-regulated phosphoprotein 21kDa
  • Antigen symbol:
    ARPP-21
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    150 µL
  • Antigen synonyms:
    RCS|TARPP|R3HDM3
  • Immunogen:
    ARPP-21
  • Purification:
    Antigen affinity purification
  • Storage buffer:
    PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
  • Storage temperature:
    Store at –20 °C
  • Cat. no.:
    10155-988
  • Supplier No.:
    11829-1-AP

Specifications

About this item

The ARPP-21 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human ARPP-21. This antibody recognizes human,mouse,rat antigen. The ARPP-21 antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 1000

Type: Primary
Antigen: ARPP-21
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Frequently Bought Together