Order Entry
ContactUsLinkComponent
Anti-BCL3 Rabbit Polyclonal Antibody
Anti-BCL3 Rabbit Polyclonal Antibody
Catalog # 10156-096
Supplier:  Proteintech
undefined
Anti-BCL3 Rabbit Polyclonal Antibody
Catalog # 10156-096
Supplier:  Proteintech
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    B-cell lymphoma 3-encoded protein
  • Antigen symbol:
    BCL3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    150 µL
  • Antigen synonyms:
    Proto-oncogene BCL3|BCL-3|D19S37|B-cell lymphoma 3 protein|BCL3|BCL4
  • Immunogen:
    BCL3
  • Purification:
    Antigen Affinity purified
  • Storage buffer:
    PBS with 0.0.2% sodium azide and 50% glycerol pH 7.3.
  • Storage temperature:
    Store at –20 °C
  • Cat. no.:
    10156-096
  • Supplier No.:
    23959-1-AP

Specifications

About this item

The BCL3 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human BCL3. This antibody recognizes human antigen. The BCL3 antibody has been validated for the following applications: ELISA, WB, IHC, IF analysis.

WB 1 : 600 ; IHC 1 : 50 ; IF 1 : 25

Type: Primary
Antigen: BCL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Frequently Bought Together