Order Entry
Canada
ContactUsLinkComponent
Anti-CAD Rabbit Polyclonal Antibody
Anti-CAD Rabbit Polyclonal Antibody
Catalog # 10154-686
Supplier:  Proteintech
undefined
Anti-CAD Rabbit Polyclonal Antibody
Catalog # 10154-686
Supplier:  Proteintech
Supplier Number:  16617-1-AP-150UL

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Carbamoyl-phosphate Synthetase 2, Aspartate transcarbamylase, And Dihydroorotase
  • Antigen symbol:
    CAD
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    150 µL
  • Antigen synonyms:
    CAD trifunctional protein|CAD protein|multifunctional protein CAD
  • Storage buffer:
    PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
  • Storage temperature:
    Store at –20 °C
  • Immunogen:
    CAD
  • Purification:
    Antigen affinity purification
  • Cat. no.:
    10154-686
  • Supplier No.:
    16617-1-AP

Specifications

About this item

The CAD Antibody from Proteintech is a rabbit polyclonal Antibody to a recombinant protein of human CAD. This Antibody recognizes human Antigen. The CAD Antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 600 ; IP 1 : 300

Type: Primary
Antigen: CAD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human