Order Entry
Canada
ContactUsLinkComponent
StabilGuard® Immunoassay Stabilizer, Surmodics IVD, Inc.
StabilGuard® Immunoassay Stabilizer, Surmodics IVD, Inc.
Catalog # 102466-310
Supplier:  Surmodics
CAS Number:  
StabilGuard® Immunoassay Stabilizer, Surmodics IVD, Inc.
Catalog # 102466-310
Supplier:  Surmodics
Supplier Number:  SG01-0050
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    StabilGuard® Immunoassay Stabilizer (BSA-Free)
  • Size:
    50 mL
  • Environmentally Preferable:
  • Protein:
    None
  • Shelf life:
    3 years
  • Storage buffer:
    PBS
  • Storage temperature:
    2 to 8° C or Room Temperature
  • pH:
    6.6-7.2
  • Cat. no.:
    102466-310
  • pH:
    6.6 - 7.2
  • Supplier No.:
    SG01-0050

Specifications

About this item

StabilGuard® Immunoassay Stabilizer (BSA-Free) effectively preserves the conformation and activity of dried proteins while simultaneously blocking the surface to reduce non-specific binding. It can be used to stabilize antibodies, antigens or enzymes on an assortment of immunoassay components including polystyrene plates, tubes, glass, membranes, and filter paper.

  • BSA/Protein Free
  • Increased Stability
  • Reduced non-specific binding
  • 3 year stability

The key benefit of the protein free formulation is the elimination of interference and cross reactivity within immunoassay applications.

StabilGuard® (BSA free) Immunoassay Stabilizers increase signal-to-noise ratios of assays through effective blocking of the plate while maximizing the binding capacity of the antigen/antibody epitopes.