MAbPac™ HIC-20 HPLC Columns, Thermo Scientific
Supplier: Thermo Scientific
Certificates
About this item
The Thermo Scientific MAbPac™ HIC-20 is designed for the separation of proteins including mAbs. This column is especially superior for the separation of mAb fragments and oxidized mAb variants.
- Optimal selectivity for mAbs and related variants
- Excellent separation for mAb fragments and oxidized variants
- High resolution and high efficiency
- Compatible with both organic solvent and aqueous mobile phase
- Rugged column stability
The MAbPac HIC-20 column is based on high-purity, spherical, wide-pore (1,000 Å), 5 μm silica particles functionalized with proprietary alkyl amide groups. The advanced surface bonding technology leads to excellent chemical stability, high resolution, and unique selectivity optimized for mAbs and different from other HIC phases.
Ordering information: Guard cartridges require use of the corresponding guard holder. See Acclaim Guard Kit Holder and Coupler V-2.
Specifications
- Catalog No:
- 10456-176
- 10436-308
- 10456-178
- Description:
- 10456-176
- 10436-308
- 10456-178
- pH range:
- 2 to 8
- 2 to 8
- 2 to 8