ContactUsLinkComponent
Hynit® 32-105 Nitrile Gloves with Slip-On Cuff, Ansell
  CA32889-902
undefined
Hynit® 32-105 Nitrile Gloves with Slip-On Cuff, Ansell
  CA32889-902
 :  3210511070
  • Glove Color:
    Blue
  • Glove size:
    7
  • Ambidextrous/Hand-specific:
    Hand-specific
  • Latex/Latex-free:
    Latex-free
  • Length:
    20.3 cm (8")
  • Environmentally Preferable:
  • EN norm:
    3111
  • Cuff style:
    Slip-on
  • Liner material:
    Cotton
  • Finish:
    Fully Coated
  • Cat. no.:
    CA32889-902
  • Supplier No.:
    3210511070
A light, comfortable and longer-lasting replacement for cotton and leather, this special nitrile formulation unites excellent resistance to abrasion.
  • A liquid-repellent impregnation repels oil, grease, and dirt
  • Excellent flexibility
Use in the automotive and transportation, building and construction, logistics and warehousing, machinery and equipment, maintenance, metal fabrication, public utilities, and recycling industries. Uses include assembly of white goods; component assembly; light material handling; packaging; shipping and receiving; steel mill general handling; and textile manufacture: threads, fabrics, mattresses, and cables.
 : ANSI level 1 cut protection; ANSI Level 3 Abrasion protection.
 : Gloves are packaged 12 pairs in a bag; 12 bags in a carton.