ContactUsLinkComponent
TBS 10X Buffer Solution, pH 7.4, Quality Biological
undefined
TBS 10X Buffer Solution, pH 7.4, Quality Biological
  • Sterility:
    Sterile filtered
  • Test description:
    DNAse, RNAse and protease tested
  • Shelf life:
    36 months
  • Shipping temperature:
    Ambient
Frequently Bought Together