ContactUsLinkComponent
WHEATON® Preassembled Open Top Caps with Septa, DWK Life Sciences
WHEATON® Preassembled Open Top Caps with Septa, DWK Life Sciences
Catalog # 66012-343
Small Business Enterprise
undefined
WHEATON® Preassembled Open Top Caps with Septa, DWK Life Sciences
Catalog # 66012-343
Supplier Number:  242245
Small Business Enterprise
Specifications
  • Cap Size:
    8-425
  • Description:
    Pre-assembled open top natural PP screw cap with septa
  • Cat. no.:
    66012-343
  • Septum Thickness:
    10 mil
  • Supplier No.:
    242245
Specifications
About this item
Natural PP cap with 10 mil septa. For use with standard opening vials (12×32 mm).