ContactUsLinkComponent
YSI MultiLab BOD/pH Lab Kits, YSI
YSI MultiLab BOD/pH Lab Kits, YSI
Supplier:  YSI
Certificates
undefined
YSI MultiLab BOD/pH Lab Kits, YSI
Supplier:  YSI
Specifications
  • Catalog No:
  • 10066-714
  • Temperature compensation:
  • Yes; Except ORP
  • Display:
  • Graphic
  • Power supply:
  • Universal
  • Memory:
  • 500 data sets in manual mode and 5,000 data sets in automatic mode
  • Interfaces:
  • Mini USB; USB-A
  • Parameters:
  • pH, ORP (mV), DO %, DO mg/L (BOD probe), Partial pressure, Conductivity, Resistivity, Salinity, TDS, Temperature, ISEs
  • Calibration:
  • DO and Conductivity = 1; pH = 1 to 5; 10 max.