Combination Lockout Starter Kit, Brady Worldwide®
CA12222-714
: Avantor
- Description:Combination Lockout Starter Kit
- Environmentally Preferable:
- Cat. no.:CA12222-714
- Supplier No.:65779
Protect employees and comply with OSHA's Lockout Standard
Starter Kit includes lockout devices to secure wall switches, plugs, or gate and ball valves. Starter kit components include; six Single Pole Breaker Lockouts, six 227 Volt "No Hole" Breaker Lockouts, six Universal Multi-Pole Breaker Lockouts, six Wall Switch Lockouts, one Small Brady Plug Lockout, one Large Brady Plug Lockout, two 1–2.5" Gate Valve Lockouts, two 2.5–5" Gate Valve Lockouts, two Small Ball Valve Lockouts, two Large Ball Valve Lockouts, five Red Labeled Lockout Hasps, six 0.75" Blue Padlocks, six 2" Blue Padlocks, twenty-five Lockout Tags, and two Yellow Canvas Locking Carrying Bags.